Order Entry
Northern Ireland
ContactUsLinkComponent
Sodium sulphide hydrate, AnalaR NORMAPUR® analytical reagent
Sodium sulphide hydrate, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Sodium sulphide hydrate, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    58.0 - 64.0 %
  • Total N (Nitrogen):
    Max. 125 ppm
  • SO3 (Sulphite):
    Max. 0.6 %
  • S2O3 (Thiosulphate):
    Max. 0.5 %

Specification Test Results

Frequently Bought Together