

Tripotassium orthophosphate monohydrate 96%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:27176-10-9
:
Formula:K₃O₄P·1H₂O
MDL Number:MFCD00150410
Melting Pt:1340 °C
:230.28 g/mol
EINECS:231-907-1
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder
- Titration with HClO4:≥95 %
- Chloride (Cl):≤50 ppm
- Sulfate (SO4):≤200 ppm
- Iron (Fe):≤20 ppm
- Heavy metals (as Pb):≤20 ppm
- Loss on ignition:≤13 % (800°C)
Specification Test Results
Frequently Bought Together