(S)-(+)-3-Chloro-1,2-propanediol 98%, ee 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:60827-45-4
:
Formula:ClCH₂CHOHCH₂OH
MDL Number:MFCD00210270
:110.54 g/mol
Boiling Pt:213 °C
Density:1,322
Flash Pt:126 °C (258 °F)
UN:2689
ADR:6.1,III
:GHS06
:GHS08
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together