Order Entry
Northern Ireland
ContactUsLinkComponent
 
Irritant
2',4',6'-Trihydroxyacetophenone monohydrate
Catalog # MOLE43063746-25G
Supplier:  Molekula
CAS Number:  
Irritant
2',4',6'-Trihydroxyacetophenone monohydrate
Catalog # MOLE43063746-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :186.16 g/mol
Boiling Pt:394 °C (1013 hPa)
Melting Pt:216…218 °C
EINECS:207-556-5
 :GHS07
Formula:C₈H₈O₄·1H₂O
MDL Number:MFCD00149091