Pesticide reference standard, 2,4'-TDE 100 µg/ml in methanol
Catalog # ACCUP-024S
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:53-19-0
- Description:2,4'-TDE 100 µg/ml in methanol
- Formula:C₁₄H₁₀Cl₄
- Description:2,4'-TDE