Pesticide reference standard, 2,4'-DDE 100 µg/ml in methanol
Catalog # ACCUP-026S
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:3424-82-6
- Description:2,4'-DDE 100 µg/ml in methanol
- Formula:C₁₄H₈Cl₄
- Description:2,4'-DDE