£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
Hydriodic acid
Hydriodic acid 47% stabilised ACS
Hydriodic acid 47% stabilised ACS
:
Thermo Fisher Scientific
Hydriodic acid 47% stabilised ACS
:
Thermo Fisher Scientific
:
10034-85-2
:
:
HI
:
MFCD00011347
:
14,4776
:
127 °C (1013 hPa)
:
1,50
:
Ambient
:
8,II
:
stab. with hypophosphorous acid 1,5%
:
127.91 g/mol
:
233-109-9
:
1787
:
GHS05
:
GHS07
msds_ALFA036490
msds_ALFA036490
Thermo Scientific Acroseal 25 Years
Frequently Bought Together
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
n-Heptane ≥99.0%, AnalaR NORMAPUR® analytical reagent
: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Petroleum spirit, 40…65 °C, anhydrous, TECHNICAL (max. 0.01% aromatic hydrocarbons)
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
: Merck
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
: Merck
Ethanol absolute ≥99.9% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Toluene, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Water USP, JP, Ph. Eur., purified
: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Dichloromethane stabilised, EMSURE® Reag. Ph. Eur., ACS, ISO for analysis, Supelco®
: Merck
Hydriodic acid 57% (w/w) in aqueous solution unstabilised for analysis, distilled
: Thermo Fisher Scientific