Aluminium copper magnesium manganese alloy (93.5:4.4:1.5:0.6 w%), alloy 2024, square bar 1.27cm (0.5in) x 1.27cm (0.5in)
Catalog # 42103.RS
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:60 cm
- Seq:7845
Specifications
About this item
:
Storage Temperature:Ambient
Formula:Al:Cu:Mg:Mn;93·5:4·4:1·5:0·6%
MDL Number:MFCD00213972