
Ethanol-[D₆], anhydrous (99% D) for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1516-08-1
:
Formula:D₃CCD₂OD
MDL Number:MFCD00051020
:52.02 g/mol
Boiling Pt:78 °C (1013 hPa)
Melting Pt:-114,5 °C
Density:0,892 g/cm³
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:216-162-2
UN:1170
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤1 % (Coulometric)
- D-Enrichment:≥98.5 %
Specification Test Results
Frequently Bought Together