

Copper ≥99.9% (metals basis), spherical powder -170+270 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-50-8
:
Formula:Cu
MDL Number:MFCD00010965
:63.55 g/mol
Boiling Pt:2567 °C (1013 hPa)
Melting Pt:1083,4 °C
Density:8,94 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-159-6
UN:3089
ADR:4.1,III
:GHS02
:GHS07
Related Information
Frequently Bought Together