Phenolphthalein 0,2% in ethanolic solution, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
CAS Number:77-09-8
:
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913
:318.33 g/mol
Density:0,884 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-004-7
UN:1170
ADR:3,II
:GHS02
:GHS07
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101)
Packaging Information
spec_VWRC31724.295
Product specification
VWR® Indicators
Product Literature
Frequently Bought Together