Order Entry
Northern Ireland
ContactUsLinkComponent
Folin-Denis' reagent for analysis of uric acid
Irritant
Folin-Denis' reagent for analysis of uric acid
Catalog # 31351.296
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Folin-Denis' reagent for analysis of uric acid
Catalog # 31351.296
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
Density:1,1 g/cm³ (20 °C)
 :GHS07
MDL Number:MFCD00146958