Order Entry
Northern Ireland
ContactUsLinkComponent
 
Allylthiourea ≥97%, TECHNICAL
Catalog # 28630.153
Supplier:  VWR Chemicals
CAS Number:  
undefined
Allylthiourea ≥97%, TECHNICAL
Catalog # 28630.153
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Assay:
    Min. 97 %

Specification Test Results

About this item

 :
 :116.19 g/mol
Merck Index:12,09500
Boiling Pt:191 °C (1013 hPa)
Melting Pt:74 °C
Density:1,219 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-683-5
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:109-57-9
Formula:H₂CCHCH₂NHC(S)NH₂
MDL Number:MFCD00004940