Order Entry
Northern Ireland
ContactUsLinkComponent
Oil 10 S, VOLTALEF®
Oil 10 S, VOLTALEF®
Catalog # 24627.188
Supplier:  VWR Chemicals
CAS Number:  
undefined
Oil 10 S, VOLTALEF®
Catalog # 24627.188
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :800 g/mol
Density:1,9 g/cm³ (20 °C)
CAS Number:9002-83-9
Formula:(C₂ClF₃)n