Order Entry
Northern Ireland
ContactUsLinkComponent
 
Ethanol denatured ≥95%, GPR RECTAPUR® (denatured with methanol)
 
undefined
Ethanol denatured ≥95%, GPR RECTAPUR® (denatured with methanol)
 

 

  • ALCH:
    94.70 - 95.10 %
  • Density (20/4):
    0.811 - 0.813 g/cm³
  • Colour number (Hazen):
    Max. 20 CO

 

Frequently Bought Together