
Succinic anhydride ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:108-30-5
:
Formula:C₄H₄O₃
MDL Number:MFCD00005525
:100.07 g/mol
UN:3261
ADR:8,III
:GHS07
Specification Test Results
- Assay:Min. 98 %
- Melting point:117 - 119 °C
Specification Test Results
Related Information
Frequently Bought Together