Hand disinfectant, DEC-HAND®
Supplier: VELTEK ASSOCIATES
Certificates
About this item
An alcohol based hand sanitizer that is processed to comply with the standards required by pharmaceutical, biotechnology, medical device, and health care industries. DEC-HAND® has multiple uses throughout any facility and complies with requirements for transport into classified areas.
- Designed for use as an instant hand sanitizer
- Packaged quadruple bagged in the ABCD Clean Room Introduction System
- Filled in ISO 5 (Grade A/B, Former Class 100)
- Filtered
- Gamma irradiated to a sterility assurance level of 10⁻⁶
- Completely validated for sterility and shelf life
The 473ml bottle can be either used with a hand pump or a wall mounted dispensing system. This product is completely traceable and has been validated for sterility and shelf life. The product is delivered with lot specific documentation.
Frequently Bought Together