About this item
SARS-CoV-2 (Severe acute respiratory syndrome coronavirus 2), also known as 2019-nCoV, is a positive-sense single-stranded RNA virus. It caused coronavirus disease 2019 (COVID-19). SARS-CoV-2 contains glycosylated spike (S) protein, which is composed of S1 subunit and S2 subunit. The S1 contains a receptor-binding domain (RBD) that can bind to ACE2 receptor on target cells.
SARS-CoV-2 Spike S1 Antibody (HC2001), Human Chimeric is produced from cell culture in vitro under conditions free from animal-derived components.
Type: Primary
Antigen: Protein
Clonality: Monoclonal
Clone: HC2001 (monoclonal antibody)
Conjugation: Unconjugated
Epitope: Confidentiality
Host: Human
Isotype: Recombinant human IgG1
Reactivity: Human
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- GENSA02038-1
- GENSA02038-100
- Antigen name:
- SARS-CoV-2 Spike Protein (RBD)
- SARS-CoV-2 Spike Protein (RBD)
- Antigen synonyms:
- Receptor-binding domain/RBD
- Receptor-binding domain/RBD
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- HC2001
- HC2001
- Reactivity:
- Human
- Human
- Host:
- Human
- Human
- Gene ID:
- MN908947
- MN908947
- Isotype:
- IgG1
- IgG1
- ELISA:
- Yes
- Yes
- Antibody fragment:
- Whole Molecule
- Whole Molecule
- Modification:
- Humanization
- Humanization
- Epitope:
- Confidentiality
- Confidentiality
- Immunogen:
- Confidentiality
- Confidentiality
- Amino acid number:
- Confidentiality
- Confidentiality
- Sequence:
- Confidentiality
- Confidentiality
- Format:
- Purified antibody
- Purified antibody
- Form:
- liquid
- liquid
- Molecular weight:
- 150 kDa
- 150 kDa
- Purification:
- Protein A
- Protein A
- Concentration:
- 1 mg/ml, supplied in PBS, pH 7.4, containing 0.02% sodium azide.
- 1 mg/ml, supplied in PBS, pH 7.4, containing 0.02% sodium azide.
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7.4, containing 0.02% sodium azide.
- PBS, pH 7.4, containing 0.02% sodium azide.
- Storage temperature:
- 20° C
- 20° C
- Shipping temperature:
- 4° C
- 4° C