Order Entry
Northern Ireland
ContactUsLinkComponent
Hydroquinone monomethyl ether ≥99%, GPR RECTAPUR®
Irritant
Hydroquinone monomethyl ether ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Hydroquinone monomethyl ether ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Melting point:
    54 …56 °C

Specification Test Results

Frequently Bought Together