(±)-1,3-Butanediol ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:107-88-0
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00004554
:90.12 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99,5 %
- Specific gravity (20°C):1,004 - 1,007
- Free acid (as CH₃COOH) :Max. 50 ppm
- Water:Max. 0,5 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC87052.290 spec_VWRC87052.320 spec_VWRC87052.460
Product specification
Frequently Bought Together