


Toluene : 2-Propanol : Water (49,5 : 50 : 0,5) % vol., Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Catalog # HONC66160-195L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Metal drum
- Pk:1
- Seq:5531
- Pk Info:195 l
Specifications
Specification Test Results
- Toluene (GC):49.5 - 50.5 Vol.%
- 2-propanol (GC):49.0 - 50.0 Vol.%
- Water (Karl Fischer):4 - 6 mg/ml
- Non-volatile matter:Max. 0.001 %
- Aluminium (Al):Max. 0.00005 %
- Barium (Ba):Max. 0.00001 %
- Bismuth (Bi):Max. 0.00001 %
- Calcium (Ca):Max. 0.00005 %
- Cadmium (Cd):Max. 0.000005 %
- Cobalt (Co):Max. 0.000002 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000002 %
- Iron (Fe):Max. 0.00001 %
- Potassium (K):Max. 0.00005 %
- Lithium (Li):Max. 0.00001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000002 %
- Molybdenum (Mo):Max. 0.00001 %
- Sodium (Na):Max. 0.00005 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.00001 %
- Strontium (Sr):Max. 0.00001 %
- Tin (Sn):Max. 0.00001 %
- Zinc (Zn):Max. 0.00001 %
- APHA:Max. 10
Specification Test Results
About this item
:
Boiling Pt:83,2 °C (1.013 hPa)
Density:0,8266 g/cm3 (20 °C)
Flash Pt:3 °C
Storage Temperature:Ambient
UN:1993
ADR:3,II
:GHS07
:GHS08
:GHS02