
Pentafluorophenylhydrazine ≥98.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Catalog # 93742-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:198.1 g/mol
:GHS07
CAS Number:828-73-9
Formula:C₆H₃F₅N₂
MDL Number:MFCD00007574