Order Entry
Northern Ireland
ContactUsLinkComponent
 
Irritant
Tridodecylmethylammonium nitrate ≥99.0%, Selectophore™ ionophore, Supelco®
Catalog # 91664-1G
Supplier:  Merck
CAS Number:  
undefined
Irritant
Tridodecylmethylammonium nitrate ≥99.0%, Selectophore™ ionophore, Supelco®
Catalog # 91664-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :599.03 g/mol
 :M-Clarity™ quality level = MQ100
 :GHS07
CAS Number:13533-59-0
Formula:[CH₃(CH₂)₁₁]₃N(NO₃)CH₃
MDL Number:MFCD00145447