Order Entry
Northern Ireland
ContactUsLinkComponent
Tetradodecylammonium nitrate ≥99.0%, Selectophore™ ionophore, Supelco®
Flammable
Irritant
Tetradodecylammonium nitrate ≥99.0%, Selectophore™ ionophore, Supelco®
Catalog # 87252-100MG
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
Tetradodecylammonium nitrate ≥99.0%, Selectophore™ ionophore, Supelco®
Catalog # 87252-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :753.32 g/mol
ADR:4.1,III
 :M-Clarity™ quality level = MQ100
UN:1325
 :GHS02
 :GHS07
CAS Number:63893-35-6
Formula:[CH₃(CH₂)₁₀CH₂]₄N(NO₃)
MDL Number:MFCD00467971