Order Entry
Northern Ireland
ContactUsLinkComponent
ω,4-Dibromoacetophenone ≥99.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
ω,4-Dibromoacetophenone ≥99.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Catalog # 68082-5G
Supplier:  Merck
CAS Number:  
undefined
ω,4-Dibromoacetophenone ≥99.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Catalog # 68082-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
 :
 :277.94 g/mol
UN:3261
ADR:8,II
 :GHS05
CAS Number:99-73-0
Formula:C₈H₆Br₂O
MDL Number:MFCD00000200