Order Entry
Northern Ireland
ContactUsLinkComponent
3-Amino-9-ethyl carbazole (AEC) ≥97.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Health
3-Amino-9-ethyl carbazole (AEC) ≥97.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Catalog # 06696-1G
Supplier:  Merck
CAS Number:  
undefined
Health
3-Amino-9-ethyl carbazole (AEC) ≥97.0% (by HPLC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Catalog # 06696-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :210.28 g/mol
UN:2811
ADR:6.1,III
 :GHS08
CAS Number:132-32-1
Formula:C₁₄H₁₄N₂
MDL Number:MFCD00004964