

o-Cresol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:95-48-7
:
Formula:C₇H₈O
MDL Number:MFCD00002226
:108.14 g/mol
UN:3455
ADR:6.1 (8),II
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 98.5 %
- Density (20/4):1.045 - 1.047
- Melting point:29 - 33 °C
- Water:Max. 0.1 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85988.180 spec_VWRC85988.290 spec_VWRC85988.320 spec_VWRC85988.460
Product specification
Frequently Bought Together