



Isopropylzinc bromide 0.5 M in THF
Catalog # H58536.AD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
Density:0,963
UN:3399
ADR:4.3,II
:Packaged under argon in resealable ChemSeal™ bottles.
:188.373 g/mol
:GHS02
:GHS05
:GHS08
:GHS07
Storage Temperature:Refrigerator
CAS Number:77047-87-1
Formula:C₃H₇BrZn
MDL Number:MFCD06200639