Order Entry
Northern Ireland
ContactUsLinkComponent
Lewatit® VP OC 1065 ~0,5 mm (weakly basic anion exchanger), Supelco®
Lewatit® VP OC 1065 ~0,5 mm (weakly basic anion exchanger), Supelco®
Catalog # 94136-100G-F
Supplier:  Merck
CAS Number:  
Lewatit® VP OC 1065 ~0,5 mm (weakly basic anion exchanger), Supelco®
Catalog # 94136-100G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
MDL Number:MFCD05865453