Venlafaxine-D6 hydrochloride solution, certified reference material, Cerilliant®, Supelco®
Catalog # V-009-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 µg/ml in methanol (as free base)
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1062606-12-5
- Description:Venlafaxine-D6 hydrochloride solution
- Formula:C₁₇D₆H₂₁NO₂ · HCl
- Description:Venlafaxine-D6 hydrochloride solution
- Pk Info:Ampoule of 1 ml
Specifications
About this item
A new internal standard suitable for many LC/MS and GC/MS applications from forensic or clinical toxicology analysis to urine drug testing and isotope dilution methods.
Also known by the brand name Effexor®, venlafaxine is an SNRI antidepressant approved for the treatment of major depressive and general anxiety disorders.
Packaging: Snap-N-Spike®/Snap-N-Shoot®