
4-Amino-2-methylquinoline ≥99.0% (by HPLC) matrix substance for MALDI-MS, Supelco®
05851-1G
: Merck
:
- Pk:1 g
- Seq:3047
- Appearance (Color):White to Off White
- Appearance (Form):Powder or Crystals
- Purity (HPLC area %):≥ 99.0 %
- Suitability test MALDI-MS:Suitable as MALDI-Matrix material
- Carbon Content:75.92% (Theory)
- Hydrogen Content:6.37% (Theory)
- Nitrogen Content:17.71% (Theory)
- Proton NMR Spectrum:Conforms to structure
- Metal trace Analysis (ICP):Corresponds to requirements
- BARIUM (ICP):≤ 5 mg/kg
- CALCIUM (ICP):≤ 20 mg/kg
- CADMIUM (ICP):≤ 5 mg/kg
- COBALT (ICP):≤ 5 mg/kg
- CHROMIUM (ICP):≤ 5 mg/kg
- COPPER (ICP):≤ 5 mg/kg
- IRON (ICP):≤ 100 mg/kg
- POTASSIUM (ICP):≤ 20 mg/kg
- MAGNESIUM (ICP):≤ 5 mg/kg
- MANGANESE (ICP):≤ 5 mg/kg
- SODIUM (ICP):≤ 50 mg/kg
- NICKEL (ICP):≤ 5 mg/kg
- LEAD (ICP):≤ 5 mg/kg
- ZINC (ICP):≤ 5 mg/kg
:
:158.2 g/mol
:162…166 °C (lit.)
:GHS07
:6628-04-2
:C₁₀H₁₀N₂
:MFCD00006759