Accessories for Battery Versaflo™ TR-830
111-2547
: 3M
- Description:Battery attachment tool, Versaflo™ TR-838
- Pk:1
Enables attachment of TR-830 batteries to TR-802N PAPR blower motors
- Bright yellow touchpoints
- to differentiate from
- non-IS versions of
- Versaflo Product Family
- Battery features bumpers
- to help absorb impact
- Mechanical attachment
- of battery to help prevent
- battery removal and
- spark risk during use
- per IEC/UL 60079
The TR-838 Battery Attachment Tool is used to mechanically attach TR-830 batteries to the TR-802N Intrinsically Safe (IS) PAPR blower motor as required by the most current UL standard (8th edition UL 913) for Intrinsically Safe PAPRs.
: UL-certified with an intrinsically safe (IS) rating of
Division 1: IS Class I, II, III; Division 1 (includes Division 2) Groups C, D, E, F, G; T4,
under the most current standard (UL 60079, 6th Edition, 2013)
Division 1: IS Class I, II, III; Division 1 (includes Division 2) Groups C, D, E, F, G; T4,
under the most current standard (UL 60079, 6th Edition, 2013)