Nitric acid : Dipicolinic acid concentrate (17 mM : 17 mM) in water eluent concentrate for IC (10x) for Metrosep C 6, Supelco®
Supplier: Merck
Certificates
Nitric acid : Dipicolinic acid concentrate (17 mM : 17 mM) in water eluent concentrate for IC (10x) for Metrosep C 6, Supelco®
Supplier: Merck
About this item
:
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
Sodium hydroxide 30% (w/w) 40% (w/v) in aqueous solution, GPR RECTAPUR® low in nitrogen
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
N-Butylthiophosphoric triamide
Supplier: Apollo Scientific