ContactUsLinkComponent
Amberlite® IR120 Hydrogen form, ca. 50% moisture 620 - 830 μm (strongly acidic cation exchanger), ion exchange resin, Supelco®
Amberlite® IR120 Hydrogen form, ca. 50% moisture 620 - 830 μm (strongly acidic cation exchanger), ion exchange resin, Supelco®
Supplier:  Merck
Certificates
Amberlite® IR120 Hydrogen form, ca. 50% moisture 620 - 830 μm (strongly acidic cation exchanger), ion exchange resin, Supelco®
Supplier:  Merck