Amberlite® XAD-16, Pore size: 200 Å 20 - 60 mesh ion-exchange resin, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:104219-63-8
:9003-69-4
Formula:C₁₀H₁₀
MDL Number:MFCD00145831
:M-Clarity™ quality level = MQ400
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
S(-)-Proline, Sigma-Aldrich®
Supplier: Merck
VWR® Flasks, Round Bottom, with Standard Ground joint
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
D-(-)-Mannitol, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals