(±)-Hydroxybupropion-D6 solution, Certified Reference Material (CRM), Ceriliant®, Supelco®
Catalog # H-062-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 μg/ml in acetonitrile
- Pack type:Ampoule
- Pk:1 mL
- Standards:Certified reference material
- CAS No.:1184984-06-2
- Description:(±)-Hydroxybupropion-D6 solution
- Formula:C₁₃D₆H₁₂ClNO₂
- Storage temperature:−20 °C
- Description:(±)-Hydroxybupropion-D6 solution
Specifications
About this item
This stable-labeled internal standard is suitable for quantitation of hydroxybupropion levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.
(±)-Hydroxybupropion is a major bupropion metabolie in urine. Bupropion is a pharmaceutical used as an antidepressant and a smoking cessation drug sold under the trade name Wellbutrin® and Zyban®.