Diluid 100 Plus for hematology (max. 0.001% acetone), J.T.Baker®
Catalog # 3961.
Supplier: Avantor
CAS Number:
Specifications
- Pk:20 L
- Seq:5302
Specifications
Specification Test Results
- Background PLT (x10E9/L):max. 5
- Background RBC (x10E12/L):max. 0.02
- Background WBC (x10E9/L):max. 0.3
- Conductivity (mS):13.10 - 14.10
- Osmolality (mOsm/kg):255 - 265
- Performance Test:passes test
- pH at 25°C:7.40 - 7.60
- Total Bacterial Growth (CFU/100ml):max. 5
Specification Test Results
About this item
:
Storage Temperature:Refrigerator
:1