Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Testosterone-d₃ solution
Supelco® Organic Reference Standard, Testosterone-d₃ solution
Catalog # T5536-1ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Testosterone-d₃ solution
Catalog # T5536-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml in 1,2-dimethoxyethane
  • Pk:
    1 mL
  • Standards:
    Analytical standard, for drug analysis
  • CAS No.:
    77546-39-5
  • Description:
    Testosterone-d₃ solution
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H225 - H315 - H332 - H360FD
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00079586
  • M-Clarity quality level:
    MQ200
  • Flash point:
    5 °C - Closed cup

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07
Caution: P202 - P210 - P233 - P303 + P361 + P353 - P304 + P340 + P312 - P308 + P313