Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Trigonelline hydrochloride
Supelco® Organic Reference Standard, Trigonelline hydrochloride
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Trigonelline hydrochloride
Supplier:  Merck

Specifications

  • CAS No.:
    6138-41-6
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00077250
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1-Methylpyridinium-3-carboxylate hydrochloride; N-Methylnicotinic acid betaine hydrochloride
  • Molecular weight:
    173,60

Specifications

Frequently Bought Together