ContactUsLinkComponent
Sigma Pseudo™ narcotic scent marijuana formulation, Supelco®
Sigma Pseudo™ narcotic scent marijuana formulation, Supelco®
  P7309-100G
 :  Merck
 :  
Sigma Pseudo™ narcotic scent marijuana formulation, Supelco®
  P7309-100G
 :  Merck
 :  
  • Pk:
    100 g
  • Description:
    Sigma Pseudo™ Narcotic Scent Marijuana formulation
  • Description:
    Sigma Pseudo(TM) Narcotic Scent Marijuana formulation
Substitute for controlled substances in the training of narcotics detector dogs. Mimics the odor of marijuana.
Sigma pseudo™ narcotic scent marijuana formulation may be used as a surrogate in the comparative study of odor profiles of real marijuana odor vs surrogate marijuana scent using multidimensional gas chromatography coupled with mass spectrometry-olfactometry (MDGC– MS-O).
 : Storage temperature: −20 °C