About this item
Canine training aids for the detection of corpses.
Sigma pseudo™ corpse scent has been used as a test material for determining the components present in it using techniques such as a direct injection, headspace solid phase microextraction (SPME), purge and trap, and thermal desorption followed by gas chromatography coupled with mass spectrometry.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Sigma Pseudo(TM) Corpse Scent