Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, 1,3,5-Trimethoxybenzene
Supelco® TraceCERT® Organic Reference Standard, 1,3,5-Trimethoxybenzene
Catalog # 74599-1G
Supplier:  Merck
CAS Number:  
undefined
Supelco® TraceCERT® Organic Reference Standard, 1,3,5-Trimethoxybenzene
Catalog # 74599-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 g
  • Standards:
    Standard for quantitative NMR, TraceCERT®
  • CAS No.:
    621-23-8
  • Description:
    1,3,5-Trimethoxybenzene
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H302
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00008385
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    Phloroglucinol trimethyl ether
  • Melting point:
    50 - 53 °C (lit.)
  • Formula:
    C₆H₃(OCH₃)₃
  • Molecular weight:
    168,19
  • Boiling point:
    255 °C (lit.)

Specifications

About this item

  • GHS signal: GHS07
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P264 - P270 - P301 + P312 - P501