Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
2-NP-AHD, analytical standard, VETRANAL™, Supelco®
2-NP-AHD, analytical standard, VETRANAL™, Supelco®
2-NP-AHD, analytical standard, VETRANAL™, Supelco®
Catalog # 33870-10MG-R
Supplier:
Merck
CAS Number:
Expand image
2-NP-AHD, analytical standard, VETRANAL™, Supelco®
Catalog # 33870-10MG-R
Supplier:
Merck
CAS Number:
Specifications
Pk:
10 mg
Standards:
analytical standard
Description:
2-NP-AHD
M-Clarity quality level:
MQ100
Description:
2-NP-AHD
Specifications
About this item
Product Details
Product Details