

Oil red O solution stain for histology, for cytology, Sigma-Aldrich®
1.02419.0250
: Merck
:
- Pack type:Plastic bottle
- Pk:250 mL
- Seq:2590
- Pk Info:Preparation Capacity 200 slides
- Suitability for microscopy:passes test
- Lipids:bright red
- Nuclei:Blue
:
:408.51 g/mol
:0,838 g/cm3
:-11 °C
:1993
:3,II
:GHS02
:GHS07
:1
:1320-06-5
:C₂₆H₂₄N₄O
:MFCD00003898
:–11 °C