Supelco® Organic Reference Standard, Yohimbine hydrochloride
Catalog # 49768-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:65-19-0
- Description:Yohimbine hydrochloride
- Storage conditions:–20 °C
- Hazard class description:H300 + H330
- Shipping conditions:STD
- MDL No.:MFCD00012674
- M-Clarity quality level:MQ100
- Synonym(s):17-Hydroxyyohimban-16-carboxylic acid methyl ester hydrochloride
- Melting point:288 - 290 °C (dec.) (lit.)
- Molecular weight:390,90
Specifications
About this item
- GHS signal: GHS06
Caution: P260 - P264 - P270 - P271 - P284 - P304 + P340 + P310