Organic reference standard, Vanadate-molybdate reagent, for phosphate determination, p.a., Supelco®
Catalog # 94685-500ML
Supplier: Merck
CAS Number:
Specifications
- Pk:500 mL
- Description:Vanadate-molybdate reagent
- M-Clarity quality level:MQ100
- Description:Vanadate-molybdate reagent
Specifications
About this item
Vanadate-molybdate reagent can be prepared by dissolving ammonium molybdate and ammonium vanadate in distilled water along with concentrated nitric acid.
Vanadate-molybdate reagent has been used to determine phosphorous using atomic absorption spectrometer, by vanadium phosphomolybdate method from cashew apple juice samples.
Reagent, suitable for eliminating interfering effects by phosphate and silicate in the AAS determination of Ca and Mg.