Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, trans-2-Decenal, analytical standard, Supelco®
Organic reference standard, trans-2-Decenal, analytical standard, Supelco®
Catalog # 91309-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, trans-2-Decenal, analytical standard, Supelco®
Catalog # 91309-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    3913-81-3
  • Description:
    trans-2-Decenal, ≥96% (GC)
  • Formula:
    C₁₀H₁₈O
  • Storage temperature:
    2...8 °C
  • Description:
    trans-2-Decenal
  • Boiling point:
    78...80 °C/3 mm Hg (lit.)

Specifications

About this item