Order Entry
Northern Ireland
ContactUsLinkComponent
 
Organic reference standard, Thymoquinone, analytical standard, Supelco®
Catalog # 03416-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Thymoquinone, analytical standard, Supelco®
Catalog # 03416-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    490-91-5
  • Description:
    Thymoquinone, ≥98,5% (GC)
  • Melting point:
    45...47 °C (lit.)
  • Formula:
    C₁₀H₁₂O₂
  • Description:
    Thymoquinone
  • Boiling point:
    230...232 °C (lit.)

Specifications

About this item

Thymoquinone is employed in preclinical studies as neuroprotective and anticonvulsant agent.

Thymoquinone has been used as an analytical standard for the determination of thymoquinone in Nigella sativa extracts and formulations by high performance thin layer chromatography (HPTLC).

It may be used as a reference standard for the identification of thymoquinone as volatile aglycone in seasonally collected Satureja montana L.and in Juniperus oxycedrus L. by gas chromatography-mass spectrometry (GC-MS).