Order Entry
Northern Ireland
ContactUsLinkComponent
 
Organic reference standard, Sudan Red B, analytical standard, Supelco®
Catalog # 51602-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Sudan Red B, analytical standard, Supelco®
Catalog # 51602-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    3176-79-2
  • Description:
    Sudan Red B, ≥97.0% (HPLC)
  • Formula:
    C₂₄H₂₀N₄O
  • Description:
    Sudan Red B

Specifications

About this item