Organic reference standard, Steviol, analytical standard, Supelco®
19345-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:471-80-7
- Description:Steviol, ≥98,0% (HPLC)
- Formula:C₂₀H₃₀O₃
- Description:Steviol
- Form:Neat
Steviol, a diterpenoiic carboxylic alcohol is an aglycone of sweet glycosides which is accumulated in Stevia rebaudiana. ent-kaurenoic acident-KA undergoes hydroxylation reaction in presence of molecular oxygen and NADPH, to yield steviol.